Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-alanine, 3-amino-N-[(1,1-dimethylethoxy)carbonyl] | 76387-70-7 | MFCD01632072 | >97.0% | 204.22 | C8H16N2O4 | 10 G
SDP

Catalog No. 5000302632
Encompass_Preferred

Boc-D-2,3-diaminopropionic acid is a Boc-protected amino acid derivative used in peptide synthesis and small-molecule research; it is supplied as a high-purity solid suitable for laboratory applications.

  • Boc-protected amino acid for peptide and small-molecule synthesis.
  • Molecular formula C8H16N2O4, molecular weight 204.22.
  • Purity ≥97.0% for research use.
  • White to off-white solid appearance.
  • Available in multiple laboratory pack sizes, including 10 g.
  • Store solid at room temperature; in solution store at -80°C or -20°C per supplier guidance.
  • Comes with datasheet, certificate of analysis, and safety data sheet.

Catalog No. 50-003-02632 Supplier Medchemexpress LLC Supplier No. HYW01469210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-D-2,3-diaminopropionic acid is a Boc-protected amino acid derivative used in peptide synthesis and small-molecule research; it is supplied as a high-purity solid suitable for laboratory applications.

  • Boc-protected amino acid for peptide and small-molecule synthesis.
  • Molecular formula C8H16N2O4, molecular weight 204.22.
  • Purity ≥97.0% for research use.
  • White to off-white solid appearance.
  • Available in multiple laboratory pack sizes, including 10 g.
  • Store solid at room temperature; in solution store at -80°C or -20°C per supplier guidance.
  • Comes with datasheet, certificate of analysis, and safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.