Learn More
Medchemexpress LLC D-alanine-d3 | 177614-69-6 | 99.3% | 92.11 | C3H4D3NO2 | 10 MG
SDP
Catalog No. 5000297883
Encompass_Preferred
D-Alanine-d3 is the deuterium-labeled isotopologue of D-alanine supplied for research use. It is reported as a weak agonist of the inhibitory glycine receptor (GlyR) and PMBA, with an EC50 of 9 mM, and is provided at high chemical purity for use as an internal standard or tracer.
  • Deuterium-labeled D-alanine (3,3,3-d3).
  • Suitable as an internal standard for mass spectrometry and tracer studies.
  • Reported weak GlyR and PMBA agonist; EC50 = 9 mM.
  • High purity (99.3%), supports analytical applications.
  • Available in small pack sizes (for example, 10 MG) for laboratory research.
Catalog No. 50-002-97883 Supplier Medchemexpress LLC Supplier No. HY41700S10MG
D-Alanine-d3 is the deuterium-labeled isotopologue of D-alanine supplied for research use. It is reported as a weak agonist of the inhibitory glycine receptor (GlyR) and PMBA, with an EC50 of 9 mM, and is provided at high chemical purity for use as an internal standard or tracer.
  • Deuterium-labeled D-alanine (3,3,3-d3).
  • Suitable as an internal standard for mass spectrometry and tracer studies.
  • Reported weak GlyR and PMBA agonist; EC50 = 9 mM.
  • High purity (99.3%), supports analytical applications.
  • Available in small pack sizes (for example, 10 MG) for laboratory research.