Learn More
Medchemexpress LLC D-Alanine-d4 | 1202063-27-1 | 99.97% | 93.12 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY41700S110MM/1ML
Encompass_Preferred
D-Alanine-d4 is the deuterium-labeled D-Alanine. D-Alanine acts as a weak GlyR (inhibitory glycine receptor) and PMBA agonist, with an EC50 of 9 mM for GlyR. It is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-04426