Learn More
Medchemexpress LLC D-Alanine-d7 | 2483831-62-3 | 99.0% | 96.14 | 1 MG
SDP
Catalog No. 5000404613
Encompass_Preferred
D-Alanine-d7 is the deuterium labeled D-Alanine. D-Alanine is a weak GlyR (inhibitory glycine receptor) and PMBA agonist, with an EC50 of 9 mM for GlyR.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-04613 Supplier Medchemexpress LLC Supplier No. HY41700S31MG
Add to Cart
Add to Cart
D-Alanine-d7 is the deuterium labeled D-Alanine. D-Alanine is a weak GlyR (inhibitory glycine receptor) and PMBA agonist, with an EC50 of 9 mM for GlyR.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.