Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-alanine-d7 | 2483831-62-3 | MFCD06658912 | 99.0% | 96.14 | C3D7NO2 | 10 MG
SDP

Catalog No. 5000299061
Encompass_Preferred

D-Alanine-d7 is a deuterium-labeled form of D-alanine used as an internal standard and tracer for quantitative analysis by NMR, GC-MS, and LC-MS. It is reported as a weak agonist of the inhibitory glycine receptor (GlyR) and PMBA, with an EC50 of 9 mM for GlyR.

  • Deuterium-labeled internal standard for NMR, GC-MS, and LC-MS.
  • Reported weak GlyR and PMBA agonist (EC50 9 mM for GlyR).
  • High isotopic purity: 99.0%.
  • Molecular formula C3D7NO2; molecular weight 96.14.
  • Available in small pack sizes (e.g., 10 mg) as a solid powder.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-99061 Supplier Medchemexpress LLC Supplier No. HY41700S310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Alanine-d7 is a deuterium-labeled form of D-alanine used as an internal standard and tracer for quantitative analysis by NMR, GC-MS, and LC-MS. It is reported as a weak agonist of the inhibitory glycine receptor (GlyR) and PMBA, with an EC50 of 9 mM for GlyR.

  • Deuterium-labeled internal standard for NMR, GC-MS, and LC-MS.
  • Reported weak GlyR and PMBA agonist (EC50 9 mM for GlyR).
  • High isotopic purity: 99.0%.
  • Molecular formula C3D7NO2; molecular weight 96.14.
  • Available in small pack sizes (e.g., 10 mg) as a solid powder.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.