Learn More
Bioworld D-ALANINE SEPAROPOREAGARO
Catalog No. 50197826
Encompass
D-ALANINE SEPAROPORE (AGAROSE) 4B-CL: Specifications: Ligand: D-Alanine; Particle size (range): 50-175m; Molecular Weight range: 6x104 - 2x107; Matrix: Separopore 4B-CL (Agarose 4% Beads Crosslinked); pH stability: 4-9; Flow specificatio...
Catalog No. 50-197-826 Supplier Bioworld Supplier No. 201810632
D-ALANINE SEPAROPORE (AGAROSE) 4B-CL: Specifications: Ligand: D-Alanine; Particle size (range): 50-175m; Molecular Weight range: 6x104 - 2x107; Matrix: Separopore 4B-CL (Agarose 4% Beads Crosslinked); pH stability: 4-9; Flow specificatio...