Learn More
Bioworld D-ALANINE SEPAROPOREAGARO
Supplier:  Bioworld 201810631
Encompass
D-ALANINE SEPAROPORE (AGAROSE) 4B-CL: Specifications: Ligand: D-Alanine; Particle size (range): 50-175m; Molecular Weight range: 6x104 - 2x107; Matrix: Separopore 4B-CL (Agarose 4% Beads Crosslinked); pH stability: 4-9; Flow specificatio...
Catalog No. 50-197-825