Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-alanine tert-butyl ester hydrochloride | 59531-86-1 | MFCD00077087 | ≥98.0% | 181.66 | C7H16ClNO2 | 500 G
SDP

Catalog No. 5000302081
Encompass_Preferred

H-D-Ala-OtBu.HCl (D-alanine tert-butyl ester hydrochloride) is the hydrochloride salt of the D-alanine tert-butyl ester, an amino acid derivative used in peptide synthesis and biochemical research. It is supplied as a high-purity white to off-white solid intended for laboratory research applications.

  • High purity ≥98.0%.
  • White to off-white solid appearance.
  • Molecular formula C7H16ClNO2; molecular weight 181.66.
  • Available in multiple laboratory sizes up to 500 g.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years).

Catalog No. 50-003-02081 Supplier Medchemexpress LLC Supplier No. HYW016426500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H-D-Ala-OtBu.HCl (D-alanine tert-butyl ester hydrochloride) is the hydrochloride salt of the D-alanine tert-butyl ester, an amino acid derivative used in peptide synthesis and biochemical research. It is supplied as a high-purity white to off-white solid intended for laboratory research applications.

  • High purity ≥98.0%.
  • White to off-white solid appearance.
  • Molecular formula C7H16ClNO2; molecular weight 181.66.
  • Available in multiple laboratory sizes up to 500 g.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.