Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Alanine tert-butyl ester hydrochloride, 98%

Catalog No. AAH5989806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5989806 5 g
AAH5989803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5989806 Supplier Thermo Scientific Chemicals Supplier No. H5989806
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 59531-86-1 | C7H16ClNO2 | 181.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59531-86-1
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00077087
InChI Key WIQIWPPQGWGVHD-NUBCRITNSA-N
Synonym d-alanine tert-butyl ester hydrochloride, h-d-ala-otbu.hcl, r-tert-butyl 2-aminopropanoate hydrochloride, r-2-aminopropionic acid tert-butyl ester hydrochloride, tert-butyl 2r-2-aminopropanoate hydrochloride, r-alanine t-butyl ester hcl, d-alanine tert-butyl ester hcl, d-alanine t-butyl ester hydrochloride, d-alanine-otbu hcl, d-alanine, 1,1-dimethylethyl ester, hydrochloride
PubChem CID 16211919
IUPAC Name tert-butyl (2R)-2-aminopropanoate;hydrochloride
SMILES CC(C(=O)OC(C)(C)C)N.Cl

Specifications

Beilstein 5763077
Quantity 5 g
Formula Weight 181.66
Optical Rotation −1.4° (c=2 in Ethanol)
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material D-Alanine tert-butyl ester hydrochloride
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.