Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Alanine tert-Butyl Ester Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A20095G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20095G 5 g
A20091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20095G Supplier TCI America Supplier No. A20095G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 59531-86-1
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00077087
InChI Key WIQIWPPQGWGVHD-NUBCRITNSA-N
Synonym d-alanine tert-butyl ester hydrochloride, h-d-ala-otbu.hcl, r-tert-butyl 2-aminopropanoate hydrochloride, r-2-aminopropionic acid tert-butyl ester hydrochloride, tert-butyl 2r-2-aminopropanoate hydrochloride, r-alanine t-butyl ester hcl, d-alanine tert-butyl ester hcl, d-alanine t-butyl ester hydrochloride, d-alanine-otbu hcl, d-alanine, 1,1-dimethylethyl ester, hydrochloride
PubChem CID 16211919
IUPAC Name tert-butyl (2R)-2-aminopropanoate;hydrochloride
SMILES CC(C(=O)OC(C)(C)C)N.Cl

Specifications

Melting Point 208°C
Color White-Yellow
Quantity 5 g
Formula Weight 181.66
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material D-Alanine tert-Butyl Ester Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.