Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-alanyl-D-alanine | 923-16-0 | MFCD00066038 | 98.0% | 160.17 | C6H12N2O3 | 1 G
SDP

Catalog No. 5000302793
Encompass_Preferred

D-Ala-D-Ala (D-alanyl-D-alanine) is a bacterial endogenous dipeptide used as an analytical and research standard in studies of peptidoglycan biosynthesis, enzyme assays, and microbiology. It is supplied as a solid with specified solubility and storage recommendations to support reproducible experimental use.

  • High purity (98.0%) suitable for analytical applications.
  • White to off-white solid, easy to handle and weigh.
  • High aqueous solubility: 100 mg/mL with ultrasonic assistance.
  • Moderate DMSO solubility: 2 mg/mL; ultrasonic and pH adjustment recommended.
  • Stable when stored as powder at 4°C or -20°C with extended shelf life.

Catalog No. 50-003-02793 Supplier Medchemexpress LLC Supplier No. HYW0154501G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Ala-D-Ala (D-alanyl-D-alanine) is a bacterial endogenous dipeptide used as an analytical and research standard in studies of peptidoglycan biosynthesis, enzyme assays, and microbiology. It is supplied as a solid with specified solubility and storage recommendations to support reproducible experimental use.

  • High purity (98.0%) suitable for analytical applications.
  • White to off-white solid, easy to handle and weigh.
  • High aqueous solubility: 100 mg/mL with ultrasonic assistance.
  • Moderate DMSO solubility: 2 mg/mL; ultrasonic and pH adjustment recommended.
  • Stable when stored as powder at 4°C or -20°C with extended shelf life.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.