Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-alanyl-L-leucine | 67113-60-4 | MFCD00066033 | 99.1% | 202.25 g/mol | C9H18N2O3 | 5 MG
SDP

Catalog No. 5000280808
Encompass_Preferred

D-Alanyl-L-leucine is a synthetic dipeptide composed of D-alanine and L-leucine, supplied as a purified solid for research and analytical applications. It serves as a source donor of L-leucine in biochemical assays and metabolic studies.

  • High purity (99.08%).
  • Molecular formula C9H18N2O3.
  • Molecular weight 202.25 g/mol.
  • Available in small pack sizes (5 mg to 100 mg) for analytical use.
  • Intended for research and analytical applications (non-clinical).
  • Stable solid form for routine laboratory handling.

Catalog No. 50-002-80808 Supplier Medchemexpress LLC Supplier No. HYW2120295MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Alanyl-L-leucine is a synthetic dipeptide composed of D-alanine and L-leucine, supplied as a purified solid for research and analytical applications. It serves as a source donor of L-leucine in biochemical assays and metabolic studies.

  • High purity (99.08%).
  • Molecular formula C9H18N2O3.
  • Molecular weight 202.25 g/mol.
  • Available in small pack sizes (5 mg to 100 mg) for analytical use.
  • Intended for research and analytical applications (non-clinical).
  • Stable solid form for routine laboratory handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.