Learn More
Medchemexpress LLC Α-D-Allofuranose, 1,2:5,6-bis-O-(1-methylethylidene)- | 2595-05-3 | 260.29 | 25 G
SDP
Catalog No. 5000449441
Encompass_Preferred
This product, 1,2:5,6-Bis-O-(1-methylethylidene)-α-D-allofuranose, is a biochemical reagent crucial for glycobiology research. It facilitates the study of sugar structure, synthesis, biology, and evolution, encompassing carbohydrate chemistry, enzymology of glycan formation and degradation, protein-glycan recognition, and the intricate roles of glycans within biological systems.
  • Biochemical reagent for glycobiology research
  • Supports carbohydrate chemistry studies
  • Useful for enzymology of glycan formation and degradation
  • Aids in protein-glycan recognition
  • Explores the role of glycans in biological systems
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-004-49441 Supplier Medchemexpress LLC Supplier No. 25GHYW008335
This product, 1,2:5,6-Bis-O-(1-methylethylidene)-α-D-allofuranose, is a biochemical reagent crucial for glycobiology research. It facilitates the study of sugar structure, synthesis, biology, and evolution, encompassing carbohydrate chemistry, enzymology of glycan formation and degradation, protein-glycan recognition, and the intricate roles of glycans within biological systems.
  • Biochemical reagent for glycobiology research
  • Supports carbohydrate chemistry studies
  • Useful for enzymology of glycan formation and degradation
  • Aids in protein-glycan recognition
  • Explores the role of glycans in biological systems
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month