Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Alloisoleucine | 1509-35-9 | MFCD00066445 | >98.0% | C6H13NO2 | 100 G
SDP

Catalog No. 5000434841
Encompass_Preferred

D-Alloisoleucine is an isoleucine derivative used for research purposes. It functions as an ergogenic supplement, affecting anabolic hormone secretion, providing fuel during exercise, enhancing mental performance under stress, and mitigating exercise-induced muscle damage.

  • Isoleucine derivative
  • For research use only
  • Can influence anabolic hormone secretion
  • Provides fuel during exercise
  • Enhances mental performance under stress
  • Helps prevent exercise-induced muscle damage
  • Recognized as a beneficial ergogenic dietary substance
  • Purity: >98.0%
  • Appearance: white to off-white solid

Catalog No. 50-004-34841 Supplier Medchemexpress LLC Supplier No. HY78912100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Alloisoleucine is an isoleucine derivative used for research purposes. It functions as an ergogenic supplement, affecting anabolic hormone secretion, providing fuel during exercise, enhancing mental performance under stress, and mitigating exercise-induced muscle damage.

  • Isoleucine derivative
  • For research use only
  • Can influence anabolic hormone secretion
  • Provides fuel during exercise
  • Enhances mental performance under stress
  • Helps prevent exercise-induced muscle damage
  • Recognized as a beneficial ergogenic dietary substance
  • Purity: >98.0%
  • Appearance: white to off-white solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.