Learn More
Medchemexpress LLC D-Allose | 2595-97-3 | 99.85% | 250 MG
SDP
Catalog No. 5000341472
Encompass_Preferred
D-Allose is an endogenous metabolite that exhibits antitumor activity against various cancer cells. It scavenges reactive oxygen species (ROS) and reduces oxidative stress damage. D-Allose also shows anti-inflammatory and neuroprotective effects through the inhibition of the TLR4/PI3K/AKT signaling pathway. Additionally, it possesses antihypertensive, cryoprotective, and anti-osteoporotic activities.
  • Exhibits antitumor activity
  • Scavenges reactive oxygen species (ROS)
  • Reduces oxidative stress damage
  • Shows anti-inflammatory effects
  • Provides neuroprotective effects
  • Inhibits the TLR4/PI3K/AKT signaling pathway
  • Possesses antihypertensive activity
  • Possesses cryoprotective activity
  • Possesses anti-osteoporotic activity
Catalog No. 50-003-41472 Supplier Medchemexpress LLC Supplier No. 250MGHY128741
D-Allose is an endogenous metabolite that exhibits antitumor activity against various cancer cells. It scavenges reactive oxygen species (ROS) and reduces oxidative stress damage. D-Allose also shows anti-inflammatory and neuroprotective effects through the inhibition of the TLR4/PI3K/AKT signaling pathway. Additionally, it possesses antihypertensive, cryoprotective, and anti-osteoporotic activities.
  • Exhibits antitumor activity
  • Scavenges reactive oxygen species (ROS)
  • Reduces oxidative stress damage
  • Shows anti-inflammatory effects
  • Provides neuroprotective effects
  • Inhibits the TLR4/PI3K/AKT signaling pathway
  • Possesses antihypertensive activity
  • Possesses cryoprotective activity
  • Possesses anti-osteoporotic activity