Learn More
Medchemexpress LLC D-Allothreonine | 24830-94-2 | 119.12 | 250 MG
SDP
Catalog No. 5000450075
Encompass_Preferred
D-Allothreonine is the D stereoisomer of Allothreonine. This peptide lipid originates from bacteria and is also a constituent of bacterial polysaccharides. It can be specifically oxidized by D-amino acid oxidase, unlike the L configuration. This compound is suitable for research into bacterial pathogenicity, antigenic diversity, and drug resistance.
  • D stereoisomer of Allothreonine
  • Peptide lipid derived from bacteria
  • Component of bacterial polysaccharides
  • Specifically oxidized by D-amino acid oxidase
  • Useful for researching bacterial pathogenicity
  • Aids in studying antigenic diversity
  • Supports research into drug resistance
  • Solid, white to off-white appearance
  • Purity of ≥98.0%
Catalog No. 50-004-50075 Supplier Medchemexpress LLC Supplier No. 250MGHYW001959
D-Allothreonine is the D stereoisomer of Allothreonine. This peptide lipid originates from bacteria and is also a constituent of bacterial polysaccharides. It can be specifically oxidized by D-amino acid oxidase, unlike the L configuration. This compound is suitable for research into bacterial pathogenicity, antigenic diversity, and drug resistance.
  • D stereoisomer of Allothreonine
  • Peptide lipid derived from bacteria
  • Component of bacterial polysaccharides
  • Specifically oxidized by D-amino acid oxidase
  • Useful for researching bacterial pathogenicity
  • Aids in studying antigenic diversity
  • Supports research into drug resistance
  • Solid, white to off-white appearance
  • Purity of ≥98.0%