Learn More
TARGETMOL CHEMICALS INC D-alpha-Hydroxyglutaric acid d
SDP
Catalog No. 502879099
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. D-alpha-Hydroxyglutaric acid disodium [Disodium (R)-2-Hydroxyglutarate] is a competitive inhibitor of alpha-ketoglutarate-dependent dioxygenases with a Ki of 0.628 mM. Purity 98%
Catalog No. 50-287-9099 Supplier TARGETMOL CHEMICALS INC Supplier No. T682050MG
Also available in 5 mg, 10 mg, 25 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. D-alpha-Hydroxyglutaric acid disodium [Disodium (R)-2-Hydroxyglutarate] is a competitive inhibitor of alpha-ketoglutarate-dependent dioxygenases with a Ki of 0.628 mM. Purity 98%