Learn More
Medchemexpress LLC HY-100542 100mg , D-alpha-Hydroxyglutaric acid (disodium salt) Disodium (R)-2-hydroxyglutarate CAS:103404-90-6 Purity:98%
SDP
Catalog No. 501493366
Encompass_Preferred
Medchemexpress, HY-100542 100mg D-alpha-Hydroxyglutaric acid (disodium salt) Disodium (R)-2-hydroxyglutarate CAS:103404-90-6 Formula:C5H6Na2O5 Ki: 10.87±1.85 mM (α-KG) Purity:98% D-alpha-Hydroxyglutaric acid disodium salt is a weak competitive α-Ketoglutarate( α-KG )-dependent dioxygenase inhibitor with K i of 10.87±1.85 mM. K i for L-Hydroxyglutaric acid (L-2-HG) is 0.628±0.036 mM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3366 Supplier Medchemexpress LLC Supplier No. HY100542100MG
Medchemexpress, HY-100542 100mg D-alpha-Hydroxyglutaric acid (disodium salt) Disodium (R)-2-hydroxyglutarate CAS:103404-90-6 Formula:C5H6Na2O5 Ki: 10.87±1.85 mM (α-KG) Purity:98% D-alpha-Hydroxyglutaric acid disodium salt is a weak competitive α-Ketoglutarate( α-KG )-dependent dioxygenase inhibitor with K i of 10.87±1.85 mM. K i for L-Hydroxyglutaric acid (L-2-HG) is 0.628±0.036 mM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.