Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Amino acid oxidase (Immobilized) | 9000-88-8 | 25 U
SDP

Catalog No. 5000396616
Encompass_Preferred

D-Amino acid oxidase (Immobilized) is an enzyme that metabolizes exogenous D-amino acids in animals for detoxifying action. It also modulates the level of D-serine in the brain. This product is intended for research use only.

  • Metabolizes exogenous D-amino acids in animals
  • Modulates the level of D-serine in the brain
  • Appearance: Solid, Yellow to brown
  • EC Number: 1.4.3.3
  • Enzyme activity: ≥50 U/g solid

Catalog No. 50-003-96616 Supplier Medchemexpress LLC Supplier No. HYP298625U
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Amino acid oxidase (Immobilized) is an enzyme that metabolizes exogenous D-amino acids in animals for detoxifying action. It also modulates the level of D-serine in the brain. This product is intended for research use only.

  • Metabolizes exogenous D-amino acids in animals
  • Modulates the level of D-serine in the brain
  • Appearance: Solid, Yellow to brown
  • EC Number: 1.4.3.3
  • Enzyme activity: ≥50 U/g solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.