Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Amphetamine hemisulfate salt Solution, 1.0 mg/mL in Methanol, Analytical Standard, For drug analysis, MilliporeSigma™ Supelco™

Catalog No. 111017195 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7195 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7195 Supplier MilliporeSigma Supelco Supplier No. A32781ML
Add to Cart
Add to Cart

Specifications

Packaging Ampule
CAS 51-63-8
Concentration 1.0 mg/mL in methanol
Linear Formula C9H13N · 0.5 H2SO4
MDL Number MFCD00069209
Industry Type Drug Analysis
Quantity 1 mL
Formula Weight 184.25
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.