Learn More
Abcam D-AP5, NMDA glutamate site antagonist, 10MG
Supplier:  Abcam AB12000310MG
Encompass_Preferred
MW 197.13 Da, Purity >99%. Competitive NMDA receptor glutamate site antagonist. More active form of DL-AP5.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0870218