Learn More
TARGETMOL CHEMICALS INC D-AP5 10MG
SDP
Catalog No. 502880608
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. D-AP5 (D-APV) is an NMDA receptor antagonist. Purity 100%
Catalog No. 50-288-0608 Supplier TARGETMOL CHEMICALS INC Supplier No. T1093010MG
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. D-AP5 (D-APV) is an NMDA receptor antagonist. Purity 100%