Learn More
Cayman Chemical D-AP5, 79055-68-8, 10 mg
SDP
Supplier:  Cayman Chemical 1453910
Encompass_Preferred
Molecular Formula C5H12NO5P, Purity ≥95%, Formula Weight 197.1, Synonyms: 5-phosphono-D-norvaline, D-2-Amino-5-Phosphonovaleric acid, D-APV; A selective NMDA receptor antagonist (Kd = 1.4 M) that competitively inhibits the glutamate binding site of NMDA receptors; widely used to study the activity of NMDA receptors particularly in regard to researching synaptic plasticity, learning, and memory.
Catalog No. NC1368401