Learn More
Apexbio Technology LLC D-AP5(Synonyms: D-2-Amino-5-phosphonopentanoic acid, D-APV, D(-)-2-Amino-5-phosphonovaleric acid), 5mg, CAS: 79055-68-8.
SDP
Catalog No. 5000060430
Encompass_Preferred
D-AP5 (CAS 79055-68-8) is a selective antagonist of the NMDA receptor It competitively inhibits glutamate binding at the NMDA receptor site exhibiting an affinity constant (Kd) of approximately 1 4 M In animal research administration of D-AP5 leads to impairment in spatial learning tasks specifically causing deficits in sensorimotor function and spatial memory performance Due to its targeted action on NMDA receptors D-AP5 serves as an important pharmacological tool for studying glutamate-related synaptic plasticity learning and memory mechanisms
Catalog No. 50-000-60430 Supplier Apexbio Technology LLC Supplier No. B62055
D-AP5 (CAS 79055-68-8) is a selective antagonist of the NMDA receptor It competitively inhibits glutamate binding at the NMDA receptor site exhibiting an affinity constant (Kd) of approximately 1 4 M In animal research administration of D-AP5 leads to impairment in spatial learning tasks specifically causing deficits in sensorimotor function and spatial memory performance Due to its targeted action on NMDA receptors D-AP5 serves as an important pharmacological tool for studying glutamate-related synaptic plasticity learning and memory mechanisms