Learn More
Medchemexpress LLC HY-100714A 5mg Medchemexpress, D-AP5 CAS:79055-68-8 Purity:>98%
SDP
Catalog No. 501931923
Encompass_Preferred
Medchemexpress, HY-100714A 5mg D-AP5 CAS:79055-68-8 D-AP5 is a NMDA receptor antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1923 Supplier Medchemexpress LLC Supplier No. HY100714A5MG
Medchemexpress, HY-100714A 5mg D-AP5 CAS:79055-68-8 D-AP5 is a NMDA receptor antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.