Learn More
Selleck Chemical LLC D-AP5-E1246-25MG
SDP
Catalog No. 503137741
Encompass_Preferred
D-AP5 (D-APV D-2-Amino-5-phosphonovaleric acid) is a selective and competitive NMDA receptor antagonist that inhibits the glutamate binding site of NMDA receptors
Catalog No. 50-313-7741 Supplier Selleck Chemical LLC Supplier No. E124625MG
D-AP5 (D-APV D-2-Amino-5-phosphonovaleric acid) is a selective and competitive NMDA receptor antagonist that inhibits the glutamate binding site of NMDA receptors