Learn More
Abcam D-AP5, NMDA glutamate site antagonist, 1MG
Catalog No. 502584728
Encompass_Preferred
MW 197.13 Da, Purity >99%. Competitive NMDA receptor glutamate site antagonist. More active form of DL-AP5.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4728 Supplier Abcam Supplier No. AB1200031MG
MW 197.13 Da, Purity >99%. Competitive NMDA receptor glutamate site antagonist. More active form of DL-AP5.
The product is subject to the following: Abcam Restricted Use Statement