Learn More
Abcam D-AP5, NMDA glutamate site antagonist, 100MG
Catalog No. 502584729
Encompass_Preferred
MW 197.13 Da, Purity >99%. Competitive NMDA receptor glutamate site antagonist. More active form of DL-AP5.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4729 Supplier Abcam Supplier No. AB120003100MG
MW 197.13 Da, Purity >99%. Competitive NMDA receptor glutamate site antagonist. More active form of DL-AP5.
The product is subject to the following: Abcam Restricted Use Statement