Learn More
Medchemexpress LLC D-Arabinopyranose | 28697-53-2 | 25 MG
SDP
Catalog No. 5000420467
Encompass_Preferred
D-Arabinopyranose is a monosaccharide and an analytical standard intended for research and analytical applications. It has been shown to exhibit strong growth inhibition against Caenorhabditis elegans with an IC50 of 7.5 mM.
  • Analytical standard for research and analytical applications
  • Exhibits strong growth inhibition against Caenorhabditis elegans
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for techniques such as HPLC, GC, and MS
  • Reference standard supplied for assay
Catalog No. 50-004-20467 Supplier Medchemexpress LLC Supplier No. 25MGHYN7082R
D-Arabinopyranose is a monosaccharide and an analytical standard intended for research and analytical applications. It has been shown to exhibit strong growth inhibition against Caenorhabditis elegans with an IC50 of 7.5 mM.
  • Analytical standard for research and analytical applications
  • Exhibits strong growth inhibition against Caenorhabditis elegans
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for techniques such as HPLC, GC, and MS
  • Reference standard supplied for assay