Learn More
Medchemexpress LLC D-Arabinose-1-C-d | 77583-92-7 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN2353S150MG
Encompass_Preferred
D-Arabinose-1-C-d is a deuterium-labeled form of arabinose, an endogenous metabolite. This compound is intended for research use only.
  • Used as a tracer in various studies.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution has the potential to influence pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-21548