Learn More
Alkali Scientific D ( ) Arabinose, 100 Grams
SDP
Catalog No. 5000660616
Encompass_Preferred
D-( ) Arabinose, 100 Grams is a high-purity sugar used in biochemical and microbiological applications. Arabinose is commonly used as a substrate for testing microbial metabolism and for various laboratory experiments requiring carbohydrate fermentation. The 100g pack size is suitable for laboratories that need moderate quantities of this compound for their research or testing. Arabinose is used in research to evaluate the fermentation capabilities of microorganisms, as it is metabolized by certain bacteria and yeast strains. This reagent is also used in genetic studies, including bacterial transformation assays and testing for enzyme activity related to sugar metabolism.
Catalog No. 50-006-60616 Supplier Alkali Scientific Supplier No. ABI001CP
D-( ) Arabinose, 100 Grams is a high-purity sugar used in biochemical and microbiological applications. Arabinose is commonly used as a substrate for testing microbial metabolism and for various laboratory experiments requiring carbohydrate fermentation. The 100g pack size is suitable for laboratories that need moderate quantities of this compound for their research or testing. Arabinose is used in research to evaluate the fermentation capabilities of microorganisms, as it is metabolized by certain bacteria and yeast strains. This reagent is also used in genetic studies, including bacterial transformation assays and testing for enzyme activity related to sugar metabolism.