Learn More
Medchemexpress LLC D-arabinose | 10323-20-3 | 10 MM 1 ML
SDP
Catalog No. 5000421048
Encompass_Preferred
D-Arabinose is an orally active antidepressant and a growth inhibitor of C. elegans. It penetrates the blood-brain barrier, selectively interferes with the metabolism of D-ribose and D-fructose, and inhibits nematode growth. It also inhibits cell biofilm synthesis, exhibits antibacterial activity, and activates the ACSS2-PPARγ/TFEB-CRTC1 axis to induce CRTC1 transcription and antidepressant-like effects.
  • Orally active antidepressant
  • Inhibits C. elegans growth
  • Penetrates the blood-brain barrier
  • Interferes with D-ribose and D-fructose metabolism
  • Inhibits nematode growth
  • Inhibits cell biofilm synthesis
  • Exerts antibacterial activity
  • Activates ACSS2-PPARγ/TFEB-CRTC1 axis
  • Induces CRTC1 transcription
  • Improves depressive-like behavior in mouse models
  • Reverses CRTC1 and BDNF downregulation
Catalog No. 50-004-21048 Supplier Medchemexpress LLC Supplier No. HYN235310MM/1ML
D-Arabinose is an orally active antidepressant and a growth inhibitor of C. elegans. It penetrates the blood-brain barrier, selectively interferes with the metabolism of D-ribose and D-fructose, and inhibits nematode growth. It also inhibits cell biofilm synthesis, exhibits antibacterial activity, and activates the ACSS2-PPARγ/TFEB-CRTC1 axis to induce CRTC1 transcription and antidepressant-like effects.
  • Orally active antidepressant
  • Inhibits C. elegans growth
  • Penetrates the blood-brain barrier
  • Interferes with D-ribose and D-fructose metabolism
  • Inhibits nematode growth
  • Inhibits cell biofilm synthesis
  • Exerts antibacterial activity
  • Activates ACSS2-PPARγ/TFEB-CRTC1 axis
  • Induces CRTC1 transcription
  • Improves depressive-like behavior in mouse models
  • Reverses CRTC1 and BDNF downregulation