Learn More
Cayman Chemical D-Arabinose, Purity: ≥95%, CAS: 10323-20-3, 250 g
SDP
Catalog No. 502913002
Encompass_Preferred
A monosaccharide; has been found in the O-specific polysaccharide of LPS in P. maltophilia; inhibits S. oralis, F. nucleatum, and P. gingivalis biofilm formation and the activity of autoinducer 2 in V. harveyi in a reporter assay at 50 and 100 mM; reduces the growth of C. elegans (IC50 = 7.5 mM), an effect that can be reversed by D-ribose or D-fructose, but not D-glucose
Catalog No. 50-291-3002 Supplier Cayman Chemical Supplier No. 35700250 G
A monosaccharide; has been found in the O-specific polysaccharide of LPS in P. maltophilia; inhibits S. oralis, F. nucleatum, and P. gingivalis biofilm formation and the activity of autoinducer 2 in V. harveyi in a reporter assay at 50 and 100 mM; reduces the growth of C. elegans (IC50 = 7.5 mM), an effect that can be reversed by D-ribose or D-fructose, but not D-glucose