Learn More
Medchemexpress LLC D-Arabitol | 488-82-4 | 50 G
SDP
Catalog No. 5000420195
Encompass_Preferred
D-Arabitol is an orally active D-enantiomer that modulates gut microbiota, increases short-chain fatty acids, and promotes AMPK-PGC-1α-related browning of white adipose tissue. It improves weight gain, fat accumulation, insulin resistance, lipid deposition, and inflammatory responses, making it applicable to obesity-related research.
  • Modulates gut microbiota composition.
  • Increases short-chain fatty acids.
  • Promotes AMPK-PGC-1α-related browning of white adipose tissue.
  • Improves weight gain, fat accumulation, insulin resistance, lipid deposition, and inflammatory responses.
  • Serves as a sole carbon/energy source for *Bacillus methanolicus* MGA3.
  • Applicable to obesity-related research.
Catalog No. 50-004-20195 Supplier Medchemexpress LLC Supplier No. HYN368650G
D-Arabitol is an orally active D-enantiomer that modulates gut microbiota, increases short-chain fatty acids, and promotes AMPK-PGC-1α-related browning of white adipose tissue. It improves weight gain, fat accumulation, insulin resistance, lipid deposition, and inflammatory responses, making it applicable to obesity-related research.
  • Modulates gut microbiota composition.
  • Increases short-chain fatty acids.
  • Promotes AMPK-PGC-1α-related browning of white adipose tissue.
  • Improves weight gain, fat accumulation, insulin resistance, lipid deposition, and inflammatory responses.
  • Serves as a sole carbon/energy source for *Bacillus methanolicus* MGA3.
  • Applicable to obesity-related research.