Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC [D-Arg1,D-Phe5,D-Trp7,9,Leu11]-Substance P | 96736-12-8 | MFCD00076802 | 98.5% | 1516.83 Da | C79H109N19O12 | 10 MG
SDP

Catalog No. 5000248571
Encompass_Preferred

[D-Arg1,D-Phe5,D-Trp7,9,Leu11]-Substance P is a synthetic modified decapeptide derived from Substance P and supplied as a lyophilized powder for laboratory research. It is used to probe neuropeptide and chemokine receptor signaling, acting as a biased agonist that modulates G12 and JNK pathways and influences neutrophil migration and Ca2+ mobilization.

  • Modified decapeptide sequence: {d-Arg}-Pro-Lys-Pro-{d-Phe}-Gln-{d-Trp}-Phe-{d-Trp}-Leu-Leu-NH2
  • High purity (98.48%)
  • Molecular weight 1516.83 Da
  • Chemical formula C79H109N19O12
  • Supplied as lyophilized powder for research use only
  • Recommended storage: powder at -80°C for long-term stability

Catalog No. 50-002-48571 Supplier Medchemexpress LLC Supplier No. HY10354410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

[D-Arg1,D-Phe5,D-Trp7,9,Leu11]-Substance P is a synthetic modified decapeptide derived from Substance P and supplied as a lyophilized powder for laboratory research. It is used to probe neuropeptide and chemokine receptor signaling, acting as a biased agonist that modulates G12 and JNK pathways and influences neutrophil migration and Ca2+ mobilization.

  • Modified decapeptide sequence: {d-Arg}-Pro-Lys-Pro-{d-Phe}-Gln-{d-Trp}-Phe-{d-Trp}-Leu-Leu-NH2
  • High purity (98.48%)
  • Molecular weight 1516.83 Da
  • Chemical formula C79H109N19O12
  • Supplied as lyophilized powder for research use only
  • Recommended storage: powder at -80°C for long-term stability

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.