Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[D-Asp3, E-Dhb7]-Microcystin-RR Solution, 10 μg/mL in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111018099 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8099 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8099 Supplier MilliporeSigma Supelco Supplier No. 325411ML
Add to Cart
Add to Cart

Microcystin is a type of toxin which can adversely affect human health. It is released by cyanobacteria and is commonly found in fresh water sources.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 202120-08-9
Flash Point 11.0°C - Closed Cup
Linear Formula C48H73N13O12
MDL Number MFCD16652550
Synonym MCYST-AspDhbRR
Type Analytical Standard
Quantity 1 mL
Formula Weight 1024.17
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 10 μg/mL in methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.