Learn More
Enzo Life Sciences [D-Asp3]microcystin-LR (25µg)
SDP
Supplier:  Enzo Life Sciences ALX350173C025
Encompass_Preferred
Isolated from Microcystis aeruginosa. Microcystin containing a leucine (L) in position 2 and arginine (R) in position 4. Additionally, the D-MeAsp at position 3 is demethylated to Asp. Hepatotoxic. Alternative name: Microcystin-LR (desmethylated). Purity: ≥95% (HPLC). Solubility: Soluble in methanol or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8772