Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich D-Asparagine

Catalog No. 44159725G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-159-725G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-159-725G Supplier Sigma Aldrich Supplier No. 44159725G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

D-Asparagine

Specifications

CAS 7787-34-0
Molecular Formula BaI2O6 -+ H2O
Linear Formula Ba(IO3)2 -+ H2O
MDL Number MFCD00239354
Quantity 100 g
Synonym (R)-2-Aminosuccinamic acid; (R)-2-Aminosuccinic acid 4-amide; D-Aspartic acid 4-amide
Molecular Weight (g/mol) 505.15 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.