Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-asparagine monohydrate | 5794-24-1 | MFCD00149558 | 98.0% | 150.13 g/mol | C4H10N2O4 | 25 G
SDP

Catalog No. 5000302711
Encompass_Preferred

D-Asparagine monohydrate is the D-enantiomer of asparagine supplied as a monohydrate for laboratory research. It is provided as a biochemical reagent for life-science studies and assays, intended for research use only and not for human or clinical applications.

  • High purity: ≥98.0% by NMR.
  • Molecular weight: 150.13 g/mol.
  • Chemical formula: C4H10N2O4.
  • Available in small-scale and bulk package sizes.
  • Supplied with certificate of analysis and safety data sheet.
  • Store refrigerated and protect from light for stability.

Catalog No. 50-003-02711 Supplier Medchemexpress LLC Supplier No. 25GHYW017444
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Asparagine monohydrate is the D-enantiomer of asparagine supplied as a monohydrate for laboratory research. It is provided as a biochemical reagent for life-science studies and assays, intended for research use only and not for human or clinical applications.

  • High purity: ≥98.0% by NMR.
  • Molecular weight: 150.13 g/mol.
  • Chemical formula: C4H10N2O4.
  • Available in small-scale and bulk package sizes.
  • Supplied with certificate of analysis and safety data sheet.
  • Store refrigerated and protect from light for stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.