Learn More
Medchemexpress LLC D-asparagine monohydrate | 5794-24-1 | MFCD00149558 | >99.0% | 150.13 g/mol | C4H10N2O4 | 500 G
SDP
Catalog No. 5000300362
Encompass_Preferred
D-asparagine monohydrate is the monohydrated form of the D-enantiomer of asparagine, supplied as a white crystalline powder for use as a biochemical reagent in research, assay development, and synthetic applications. The substance has CAS 5794-24-1 and a molecular weight of 150.13 g/mol, and is provided with accompanying documentation for laboratory use.
  • High purity (>99%) suitable for analytical and synthetic work.
  • Provided as a stable, easy-to-handle white crystalline powder.
  • Recommended storage at 4°C and protection from light to preserve integrity.
  • Suitable for biochemical assays, method development, and standards preparation.
  • Includes product data sheet and certificate of analysis for quality assurance.
Catalog No. 50-003-00362 Supplier Medchemexpress LLC Supplier No. HYW017444500G
D-asparagine monohydrate is the monohydrated form of the D-enantiomer of asparagine, supplied as a white crystalline powder for use as a biochemical reagent in research, assay development, and synthetic applications. The substance has CAS 5794-24-1 and a molecular weight of 150.13 g/mol, and is provided with accompanying documentation for laboratory use.
  • High purity (>99%) suitable for analytical and synthetic work.
  • Provided as a stable, easy-to-handle white crystalline powder.
  • Recommended storage at 4°C and protection from light to preserve integrity.
  • Suitable for biochemical assays, method development, and standards preparation.
  • Includes product data sheet and certificate of analysis for quality assurance.