Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC d-Atabrine dihydrochloride | 56100-41-5 | 99.6% | 472.88 | 1 ML
SDP

Catalog No. 5000351316
Encompass_Preferred

d-Atabrine dihydrochloride is an active enantiomer of quinacrine which displays antiprion activity.

  • Purity of 99.64%
  • Molecular weight of 472.88
  • Appears as a solid
  • Color is light yellow to yellow
  • Recommended storage at 4°C, sealed, away from moisture
  • For solutions, store at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture

Catalog No. 50-003-51316 Supplier Medchemexpress LLC Supplier No. HY13735D10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

d-Atabrine dihydrochloride is an active enantiomer of quinacrine which displays antiprion activity.

  • Purity of 99.64%
  • Molecular weight of 472.88
  • Appears as a solid
  • Color is light yellow to yellow
  • Recommended storage at 4°C, sealed, away from moisture
  • For solutions, store at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.