Learn More
Selleck Chemical LLC d-Bicuculline
SDP
Catalog No. 5000002098
Encompass_Preferred
( )-Bicuculline (d-Bicuculline) is a competitive antagonist of GABAA receptors with IC50 of 2 M also blocks Ca(2 )-activated potassium channels
Catalog No. 50-000-02098 Supplier Selleck Chemical LLC Supplier No. S707110MM/1ML
( )-Bicuculline (d-Bicuculline) is a competitive antagonist of GABAA receptors with IC50 of 2 M also blocks Ca(2 )-activated potassium channels