Learn More
Enzo Life Sciences D-(+)-Biotin (1g). CAS: 58-85-5
SDP
Catalog No. 502009282
Encompass_Preferred
Alternative name: Vitamin H. Purity: ≥98% (HPLC). Solubility: Soluble in hot water, diluted acids and alkaline solutions; hardly soluble in cold water. Long Term Storage: +4°C.
Catalog No. 50-200-9282 Supplier Enzo Life Sciences Supplier No. ALX460002G001
Alternative name: Vitamin H. Purity: ≥98% (HPLC). Solubility: Soluble in hot water, diluted acids and alkaline solutions; hardly soluble in cold water. Long Term Storage: +4°C.