Learn More
AdipoGen D-Biotin, 58-85-5, MFCD00005541, 1 g
SDP
Catalog No. 501146946
Encompass_Preferred
MF: C10H16N2O3S, MW: 244.3, ≥98%, Appearance: White to off-white powder. D-(+)-Biotin, also known as vitamin H or coenzyme R, is a water-soluble B-vitamin (vitamin B7).
Catalog No. 50-114-6946 Supplier AdipoGen Supplier No. CDXB0148G001
MF: C10H16N2O3S, MW: 244.3, ≥98%, Appearance: White to off-white powder. D-(+)-Biotin, also known as vitamin H or coenzyme R, is a water-soluble B-vitamin (vitamin B7).