Learn More
Gold Biotechnology Inc D Biotin 5 g
SDP
Catalog No. 5000562060
Encompass_Preferred
Biotin can be used in cell culture media for its involvement in a number of cellular processes including gluconeogenesis synthesis of fatty acids and amino acids isoleucine and valine
Catalog No. 50-005-62060 Supplier Gold Biotechnology Inc Supplier No. B9505
Biotin can be used in cell culture media for its involvement in a number of cellular processes including gluconeogenesis synthesis of fatty acids and amino acids isoleucine and valine