Learn More
Bioworld D-Biotin, Vitamin H, 25 g
Catalog No. 50255399
Encompass_Preferred
Used as a vitamin in culture media and biotenylation of proteins and nucleic acids. Technical Information:Reagent GradeAppearance: White Crystalline NeedlesDensity : 1.267 g/cm3HS Code : 29362930IUPAC_Name: 5-[(3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl]pentanoic acidMelting Point: 231-233 °CRTECS Number: XJ9088200
Catalog No. 50-255-399 Supplier Bioworld Supplier No. 402000804
Used as a vitamin in culture media and biotenylation of proteins and nucleic acids. Technical Information:Reagent GradeAppearance: White Crystalline NeedlesDensity : 1.267 g/cm3HS Code : 29362930IUPAC_Name: 5-[(3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl]pentanoic acidMelting Point: 231-233 °CRTECS Number: XJ9088200