Learn More
Alkali Scientific D (+) Biotin [Vitamin H], 500 Mg
SDP
Catalog No. 5000660348
Encompass_Preferred
D (+) Biotin [Vitamin H], 500mg, is a laboratory-grade reagent used in biochemical and cellular research. Biotin, also known as Vitamin H, is a water-soluble B-vitamin essential for cell growth, fatty acid synthesis, and the metabolism of amino acids and carbohydrates. This reagent is commonly used in the preparation of cell culture media, particularly for research involving biotinylation of proteins, nucleic acids, or lipids. Biotin is also crucial in metabolic processes and is involved in gene expression, cellular growth, and signaling pathways. Laboratories use this high-quality reagent to ensure optimal biotin levels in experiments, ensuring reproducibility and accuracy in biochemical studies and research.
Catalog No. 50-006-60348 Supplier Alkali Scientific Supplier No. DBI001CP
D (+) Biotin [Vitamin H], 500mg, is a laboratory-grade reagent used in biochemical and cellular research. Biotin, also known as Vitamin H, is a water-soluble B-vitamin essential for cell growth, fatty acid synthesis, and the metabolism of amino acids and carbohydrates. This reagent is commonly used in the preparation of cell culture media, particularly for research involving biotinylation of proteins, nucleic acids, or lipids. Biotin is also crucial in metabolic processes and is involved in gene expression, cellular growth, and signaling pathways. Laboratories use this high-quality reagent to ensure optimal biotin levels in experiments, ensuring reproducibility and accuracy in biochemical studies and research.