Learn More
Medchemexpress LLC D-Biotinol 25mg
SDP
Catalog No. 5000206228
Encompass_Preferred
D-Biotinol is the nutrition of Lactobacillus arabinosus L casei or Saccharomyces cerevisiae D-Biotinol replaces the D-biotin (HY-B0511) in saving egg white induced biotin deficiency in rats D-Biotinol is orally active and displays to be converted to biotin by rats[1]
Catalog No. 50-002-06228 Supplier Medchemexpress LLC Supplier No. 25MGHYW096159
D-Biotinol is the nutrition of Lactobacillus arabinosus L casei or Saccharomyces cerevisiae D-Biotinol replaces the D-biotin (HY-B0511) in saving egg white induced biotin deficiency in rats D-Biotinol is orally active and displays to be converted to biotin by rats[1]