Learn More
Lonza Walkersville D-BSMC-As - Asthma
Catalog No. 5000496263
Encompass
Human bronchial smooth muscle cells are isolated from the major bronchia of asthmatic donors Cryopreserved Diseased Bronchial Smooth Muscle cells stain positive for alpha smooth muscle actin and negative for von Willebrandfactor VIII All cells test negative for mycoplasma bacteria yeast and fungi HIV-1 hepatitis B and hepatitisC are not detected for all donors and/or cell lots A Certificate of Analysisis provided for each cell lot purchased
Catalog No. 50-004-96263 Supplier Lonza Walkersville Supplier No. 00194850
Human bronchial smooth muscle cells are isolated from the major bronchia of asthmatic donors Cryopreserved Diseased Bronchial Smooth Muscle cells stain positive for alpha smooth muscle actin and negative for von Willebrandfactor VIII All cells test negative for mycoplasma bacteria yeast and fungi HIV-1 hepatitis B and hepatitisC are not detected for all donors and/or cell lots A Certificate of Analysisis provided for each cell lot purchased