Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Camphor, Analytical Standard, ≥97.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019360 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9360 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9360 Supplier MilliporeSigma Supelco Supplier No. 50843100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Camphor is an acyclic terpenoid, that occurs naturally in the wood of Cinnamomum camphora.

TRUSTED_SUSTAINABILITY

Specifications

CAS 464-49-3
Vapor Pressure 4 mmHg (70°C)
Vapor Density 5.24 (vs. air)
Physical Form Neat
Linear Formula C10H16O
MDL Number MFCD00064149
Melting Point 178°C to 182°C
Synonym 2-Bornanone; 2-Camphanone
Quantity 100 mg
Formula Weight 152.23
Percent Purity ≥97.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.